Anti-IgG3 Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
1102-09
100242-760EA 468.15 USD
100242-760
Anti-IgG3 Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat F(ab’)2 Anti-Mouse IgG3-PE antibody with minimal reactivity to human proteins for use in flow cytometry assays.

Type: Secondary
Antigen: IgG3
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR