Anti-IgG3 Goat Antibody

Supplier: Southern Biotechnology
1102-01
100242-754EA 285.08 USD
100242-754
Anti-IgG3 Goat Antibody
Antibodies
Purified Goat F(ab’)2 Anti-Mouse IgG3 antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, and immunocytochemistry assays.

Type: Secondary
Antigen: IgG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR