Anti-IgG3 Goat Antibody (Alexa Fluor® 555)

Supplier: Southern Biotechnology
1100-32
103259-832EA 381.85 USD
103259-832
Anti-IgG3 Goat Antibody (Alexa Fluor® 555)
Antibodies
Goat Anti-Mouse IgG3-Alexa Fluor 555 antibody with minimal reactivity to human proteins for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: IgG3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR