Anti-IgG2b Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
1093-02
103259-818EA 299.46 USD
103259-818
Anti-IgG2b Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-Mouse IgG2b-FITC antibody with minimal reactivity to human, bovine, and horse serum proteins for use in western blotting, immunocytochemistry, and other fluorescent assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR