Anti-IgG2b Goat Antibody

Supplier: Southern Biotechnology
1092-01
100242-726EA 285.08 USD
100242-726
Anti-IgG2b Goat Antibody
Antibodies
Purified Goat F(ab’)2 Anti-Mouse IgG2b antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, and immunocytochemistry assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR