Anti-IgG2b Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
1091-05
103259-812EA 137.31 USD
103259-812
Anti-IgG2b Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Mouse IgG2b-HRP antibody for use in ELISA assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR