Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Mouse IgG2b Antibody (Alexa Fluor® 555)

Supplier: SOUTHERN BIOTECHNOLOGY MS
1090-32
103259-804
Goat Anti-Mouse IgG2b Antibody (Alexa Fluor® 555)
Goat Anti-Mouse IgG2b-Alexa Fluor 555 antibody with minimal reactivity to human proteins for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty