Anti-IgG2b Goat Antibody (Alexa Fluor® 647)

Supplier: Southern Biotechnology
1090-31
103259-802EA 381.85 USD
103259-802
Anti-IgG2b Goat Antibody (Alexa Fluor® 647)
Antibodies
Goat Anti-Mouse IgG2b-Alexa Fluor 647 antibody with minimal reactivity to human proteins for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR