Anti-IgG2a Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
1083-05
103259-796EA 166.08 USD
103259-796
Anti-IgG2a Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Mouse IgG2a-HRP antibody with minimal reactivity to human, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR