Anti-IgG2a Goat Antibody (AP (Alkaline Phosphatase))

Supplier: Southern Biotechnology
1083-04
103259-794EA 187 USD
103259-794
Anti-IgG2a Goat Antibody (AP (Alkaline Phosphatase))
Antibodies
Goat Anti-Mouse IgG2a-AP antibody with minimal reactivity to human, bovine, and horse serum proteins for use in ELISA assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR