Anti-IgG2a Goat Antibody (AP (Alkaline Phosphatase))

Supplier: Southern Biotechnology
1081-04
103259-786EA 151.69 USD
103259-786
Anti-IgG2a Goat Antibody (AP (Alkaline Phosphatase))
Antibodies
Goat Anti-Mouse IgG2a-AP antibody for use in ELISA, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR