Anti-IgG2a Goat Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Southern Biotechnology
1080-03
100242-676EA 287.69 USD
100242-676
Anti-IgG2a Goat Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Goat Anti-Mouse IgG2a-TRITC antibody with minimal reactivity to human proteins for use in immunohistochemistry / immunocytochemistry assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR