Anti-IgG2a Goat Antibody

Supplier: Southern Biotechnology
1080-01
100242-672EA 258.92 USD
100242-672
Anti-IgG2a Goat Antibody
Antibodies
Purified Goat Anti-Mouse IgG2a antibody with minimal reactivity to human proteins for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, ELISpot, and purification assays.

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR