Anti-Mouse Lambda Goat Antibody (RPE (R-Phycoerythrin))

Supplier: Southern Biotechnology
1060-09
100242-612EA 388.38 USD
100242-612
Anti-Mouse Lambda Goat Antibody (RPE (R-Phycoerythrin))
Antibodies
Goat Anti-Mouse Lambda-PE antibody for use in flow cytometry assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR