Anti-Mouse Lambda Goat Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Southern Biotechnology
1060-02
100242-600EA 281.15 USD
100242-600
Anti-Mouse Lambda Goat Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat Anti-Mouse Lambda-FITC antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR