Anti-Mouse Lambda Goat Antibody

Supplier: Southern Biotechnology
1060-01
100242-598EA 238 USD
100242-598
Anti-Mouse Lambda Goat Antibody
Antibodies
Purified Goat Anti-Mouse Lambda antibody for use in ELISA, flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, immunoprecipitation, ELISpot, stimulation, and separation assays.

Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR