Anti-Mouse Kappa Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Southern Biotechnology
1050-05
100242-580EA 252.38 USD
100242-580
Anti-Mouse Kappa Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat Anti-Mouse Kappa-HRP antibody for use in ELISA, immunohistochemistry / immunocytochemistry, western blotting, and ELISpot assays.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR