HOBO Temperature, Relative Humidity, Light, External Data Logger, Onset Computer Corp
U12-012
MSPP-U12-012EA 89.6 USD
MSPP-U12-012
HOBO Temperature, Relative Humidity, Light, External Data Logger, Onset Computer Corp
The data logger measure temperature, relative humidity and light intensity with 12-bit resolution measurements and gathers a range of recorded data and stores 43000 measurements.
  • Provides high accuracy
  • Operates in an indoor environment
  • Large memory for long-term deployments or fast sampling
  • Programmable and push button start
The data logger supports the measuremnts such as 4 - 20 mA, AC current, AC voltage, air velocity, carbon dioxide, compressed air flow, DC current, DC voltage, gauge pressure, kilowatts (kW), light intensity, relative humidity, temperature and volatile organic compund.
Compatible with Onset’s HOBO U-Shuttle for convenient data transport.
Compatible with HOBOware and HOBOware Pro for logger setup, graphing and analysis.
Certifications: CE marked.