Affinity Removal Spin Cartridge HSA, Agilent Technologies
Supplier: Agilent Technologies
5188-5334 5188-5334
97066-652EA 784.07 USD
97066-652 CA97066-652
Affinity Removal Spin Cartridge HSA, Agilent Technologies
Protein Purification Protein Purification Kits
The affinity removal spin cartridge – human albumin is designed to remove the most interfering high-abundant protein (albumin) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and luer-lok syringes
  • Serum load volume is 65 µl per run
  • Optimize sample recovery by utilizing our rapid, single injection protocols and optimized buffers
Removal of albumin improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.