SOUT0430
0430-01
101442-580EA 52 USD
101442-580
SOUT0430
BCIP/NBT Membrane Substrate for western blotting produces an insoluble, dark blue end product.
  • 5-Bromo-4-chloro-3-indolyl phosphate/nitro blue tetrazolium for western blotting
  • Produces an insoluble, dark blue end product
  • Ready-to-use, one component formulation
BCIP/NBT Membrane Substrate for western blotting is ready to use and produces an insoluble, dark blue end product upon reaction with alkaline phosphatase (AP).