SOUT0404
0404-01
103265-568EA 42 USD
103265-568
SOUT0404
ABTS One Component Microwell ELISA Substrate is ready to use and produces a soluble, blue-green end product upon reaction with horseradish peroxidase (HRP) that is measurable between 405 nm and 410 nm.
  • Ready-to-use, one component formulation
  • Measurable between 405 nm and 410 nm
Product Use
Ready to use formulation; dilution of the substrate is not recommended
Reaction may be stopped with 1% sodium dodecyl sulfate (SDS)