CP-547632 ≥98% (by HPLC)

Synonyms: PAN908063-((4-bromo-2,6-difluorobenzyl)oxy)-5-(3-(4-(pyrrolidin-1-yl)butyl)ureido)isothiazole-4-carboxamide

B2056-1000 B2056-500
76269-520EA 110.91 USD
76269-520 76269-522
CP-547632 ≥98% (by HPLC)
CP-547632

CP-547,632 is a potent inhibitor of the receptor tyrosine kinases VEGF receptor 2 (VEGFR2) and basic fibroblast growth factor (bFGF) with IC₅₀ values of 11 and 9 nM, respectively. Displays anticancer activity.

Formula: C₂₀H₂₄BrF₂N₅O₃S
MW: 532,4 g/mol
CAS Number: 252003-65-9
https://us.vwr.com/stibo/web/std.lang.all/03/40/24140340.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR