BMS-794833 ≥98% (by HPLC)

Danger

Synonyms: N-(4-((2-amino-3-chloropyridin-4-yl)oxy)-3-fluorophenyl)-5-(4-fluorophenyl)-4-oxo-1,4-dihydropyridine-3-carboxamide

B2055-5 B2055-25
76269-516EA 157.61 USD
76269-516 76269-518
BMS-794833 ≥98% (by HPLC)
BMS-794833

BMS-794833 is a potent, ATP-competitive inhibitor of Met and VEGFR2 with IC₅₀ of 1.7 nM and 15 nM, respectively. It also inhibits Ron, Axl, and Flt-3 with IC₅₀ values <3 nM.

Formula: C₂₃H₁₅ClF₂N₄O₃
MW: 468,84 g/mol
MDL Number: MFCD18206786
CAS Number: 1174046-72-0
https://us.vwr.com/stibo/web/std.lang.all/03/39/24140339.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR