Topotecan hydrochloride ≥98% (by HPLC)

Danger

Synonyms: 10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione hydrochloride

B2050-50 B2050-10
76269-496EA 88.63 USD
76269-496 76269-498
Topotecan hydrochloride ≥98% (by HPLC)
Topotecan hydrochloride

Topocecan is a semisynthetic derivative of Camptothecin (Cat.No.1039) that displays antineoplastic activity. DNA topoisomerase I inhibitor; induces rapid apoptotic death in human B-lineage acute lymphoblastic leukemia (ALL) cells. Improves survival in three mouse models of poor prognosis.

Formula: C₂₃H₂₃N₃O₅.HCl
MW: 457,91 g/mol
CAS Number: 119413-54-6
https://us.vwr.com/stibo/web/std.lang.all/03/34/24140334.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR