Cyclic-di-GMP ≥95%

Synonyms: Guanylyl-(3'→5')-3'-guanylic acidc-di-GMPCyclic diguanylate3',5'-Cyclic diguanylic acidcyclic nucleotide

B2045-250 B2045-1000
76269-314EA 170.52 USD
76269-314 76269-316
Cyclic-di-GMP ≥95%
Cyclic-di-GMP

Cyclic di-GMP (c-di-GMP) is one of the most important and common bacterial second messenger. It is involved in numerous prokaryotic processes, including biofilm formation, motility, virulence, and cell cycling. c-di-GMP also has functions in eukaryotic cells. It is detected by the transmembrane protein stimulator of interferon genes (STING), leading to activation of the innate immune system.

Formula: C₂₀H₂₄N₁₀O₁₄P₂
MW: 690,41 g/mol
CAS Number: 61093-23-0
https://us.vwr.com/stibo/web/std.lang.all/03/29/24140329.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR