Maraviroc ≥98% (by HPLC)

Synonyms: 4,4-Difluoro-N-((S)-3-((1R,3R,5S)-3-(3-isopropyl-5-methyl-4H-1,2,4-triazol-4-yl)-8-azabicyclo[3.2.1]octan-8-yl)-1-phenylpropyl)cyclohexane-1-carboxamideUK4278574,4-difluoro-N-[(1S)-3-[(3-exo)-3-[3-methyl-5-(1-methylethyl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]oct-8-yl]-1-phenylpropyl]-cyclohexanecarboxamideMVC

B2044-5 B2044-25
76269-310EA 145.93 USD
76269-310 76269-312
Maraviroc ≥98% (by HPLC)
Maraviroc

Maraviroc is a potent and selective C-C chemokine receptor type 5 (CCR5) antagonist drug that is designed to act against HIV by interfering with the interaction between HIV and CCR5.

Formula: C₂₉H₄₁F₂N₅O
MW: 513,67 g/mol
MDL Number: MFCD13188530
CAS Number: 376348-65-1
https://us.vwr.com/stibo/web/std.lang.all/03/28/24140328.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR