6-Mercaptopurine monohydrate

Danger

Synonyms: 6-Thiohypoxanthine monohydrate7H-Purine-6-thiol hydrate6-Purinethiol monohydrate

B2039-250 B2039-50
76269-290EA 75.89 USD
76269-290 76269-292
6-Mercaptopurine monohydrate
6-Mercaptopurine monohydrate

6-Mercaptopurine is an inhibitor of de novo purine synthesis through interference with DNA and RNA synthesis. An immunosuppressive and antileukemic agent. 

Formula: C₅H₄N₄S·H₂O
MW: 170.19 g/mol (152.17 anhy)
MDL Number: MFCD03854445
CAS Number: 6112-76-1
https://us.vwr.com/stibo/web/std.lang.all/03/17/24140317.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR