ZLN024 hydrochloride ≥98% (by HPLC)

Synonyms:  2-[[2-(2-Bromo-4-methylphenoxy)ethyl]thio]pyrimidine hydrochloride

B2034-5 B2034-25
76269-466EA 145.93 USD
76269-466 76269-468
ZLN024 hydrochloride ≥98% (by HPLC)
ZLN024 hydrochloride

ZLN024 is an allosteric activator of AMP-activated protein kinase (AMPK) heterotrimers (EC₅₀s = 0.42 and 0.95 µM for α1β1γ1 and α2β1γ1, respectively. Inhibits dephosphorylation of AMPK Thr-172 by PP2Cα. Stimulates glucose uptake and fatty acid oxidation without increasing the ADP/ATP ratio.

Formula: C₁₃H₁₃BrN₂OS.HCl
MW: 361,69 g/mol
CAS Number: 723249-01-2
https://us.vwr.com/stibo/web/std.lang.all/03/11/24140311.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR