GF-627368X ≥98% (by HPLC)

Synonyms: 2-(4-(4,9-Diethoxy-1-oxo-1H-benzo[f]isoindol-2(3H)-yl)phenyl)-N-(phenylsulfonyl)acetamide4-(4-,9-Diethoxy-1,3-dihydro-1-oxo-2H-benz[f]isoindol-2-yl)-N-(phenylsulfonyl)benzeneacetamide4-(4-,9-Diethoxy-1,3-dihydro-1-oxo-2H-benz[f]isoindol-2-yl)-N-(phenylsulphonyl)benzeneacetamide2-(4-(4,9-Diethoxy-1-oxo-1H-benzo[f]isoindol-2(3H)-yl)phenyl)-N-(phenylsulphonyl)acetamide

B2029-1
76269-448EA 145.93 USD
76269-448
GF-627368X ≥98% (by HPLC)
GF-627368X

GW-627368X is a potent and selective competitive antagonist of the prostanoid EP4 receptor with additional affinity at TP receptors (pKi values are 7.0 and 6.8 in competition radioligand bioassays). Displays antitumor activity.

Formula: C₃₀H₂₈N₂O₆S
MW: 544,62 g/mol
MDL Number: MFCD12912327
CAS Number: 439288-66-1
https://us.vwr.com/stibo/web/std.lang.all/03/06/24140306.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR