Deflazacort

B2023-50 B2023-250
76269-424EA 87.57 USD
76269-424 76269-426
Deflazacort
Deflazacort

Deflazacort is a glucocorticoid that displays anti-inflammatory and immunosuppressive properties.

Formula: C₂₅H₃₁NO₆
MDL Number: MFCD00866106
CAS Number: 14484-47-0
https://us.vwr.com/stibo/web/std.lang.all/03/00/24140300.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR