ACTB-1003 ≥98%

Synonyms: 1-(4-(4-amino-6-(methoxymethyl)-7-(morpholinomethyl)pyrrolo[2,1-f][1,2,4]triazin-5-yl)-2-fluorophenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea

B2016-5 B2016-25
76269-396EA 206.79 USD
76269-396 76269-398
ACTB-1003 ≥98%
ACTB-1003

ACTB-1003 is a potent kinase inhibitor with IC₅₀s of 6, 2 and 4 nM for FGFR1, VEGFR2 and Tie-2, respectively. Displays anticancer activities.

Formula: C₂₇H₂₆F₅N₇O₃
MW: 591,53 g/mol
CAS Number: 939805-30-8
https://us.vwr.com/stibo/web/std.lang.all/02/93/24140293.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR