Maytansinol ≥95%

Synonyms: (14S,16S,32S,33S,2R,4S,10E,12E,14R)-86-chloro-14,4-dihydroxy-85,14-dimethoxy-33,2,7,10-tetramethyl-7-aza-1(6,4)-oxazinana-3(2,3)-oxirana-8(1,3)-benzenacyclotetradecaphane-10,12-diene-12,6-dioneAnsamitocin P-0

B2015-5 B2015-25
76269-392EA 157.61 USD
76269-392 76269-394
Maytansinol ≥95%
Maytansinol

Maytansinol is a potent microtubule depolymerizing agent. It inhibits the polymerization of tubulin and depolymerizes the once polymerized tubulin.

Formula: C₂₈H₃₇ClN₂O₈
MW: 565,06 g/mol
MDL Number: MFCD28144515
CAS Number: 57103-68-1
https://us.vwr.com/stibo/web/std.lang.all/02/92/24140292.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR