AX-024 hydrochloride ≥98% (by HPLC)

Synonyms: AX0241-((4-(4-Fluorophenyl)-6-methoxy-2H-chromen-3-yl)methyl)pyrrolidine hydrochloride

B2013-5 B2013-25
76269-336EA 169.28 USD
76269-336 76269-338
AX-024 hydrochloride ≥98% (by HPLC)
AX-024 hydrochloride

AX-024 is a T cell receptor (TCR) inhibitor. An orally available inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation (IC₅₀ = 1 nM). By modulating TCR signaling, the inhibitor prevents the development of psoriasis and asthma. Additionally, it exerts a long-lasting therapeutic effect in a model of autoimmune encephalomyelitis.

Formula: C₂₁H₂₂FNO₂.HCl
MW: 375,86 g/mol
MDL Number: MFCD31628548
CAS Number: 1704801-24-0
https://us.vwr.com/stibo/web/std.lang.all/02/90/24140290.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR