CCF-642 ≥98% (by HPLC)

Warning

Synonyms: 3-(4-Methoxyphenyl)-5-[(5-nitro-2-thienyl)methylene]-2-thioxo-4-thiazolidinone

B2009-25 B2009-5
76269-278EA 145.93 USD
76269-278 76269-280
CCF-642 ≥98% (by HPLC)
CCF-642

CCF-642 is a protein disulfide isomerase (PDI) inhibitor that displays anticancer activity in multiple myeloma. Consistent with PDI inhibition, CCF642 causes acute ER stress in multiple myeloma cells accompanied by apoptosis-inducing calcium release. 

Formula: C₁₅H₁₀N₂O₄S₃ 
MW: 378,44 g/mol
CAS Number: 346640-08-2 
https://us.vwr.com/stibo/web/std.lang.all/02/86/24140286.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR