QNZ46 ≥98% (by HPLC)

Synonyms: 4-[6-Methoxy-2-[(1E)-2-(3-nitrophenyl)ethenyl]-4-oxo-3(4H)-quinazolinyl]benzoic acid(E)-4-(6-Methoxy-2-(3-nitrostyryl)-4-oxoquinazolin-3(4H)-yl)benzoic acid

B2007-5 B2007-25
76269-384EA 145.93 USD
76269-384 76269-386
QNZ46 ≥98% (by HPLC)
QNZ46

QNZ46 is a NR2C/NR2D-selective NMDA receptor non-competitive antagonist (IC₅₀ values are 3, 6, 229, and >300, >300 μM for NR2D, NR2C, NR2A, NR2B, and GluR1, respectively)

Formula: C₂₄H₁₇N₃O₆ 
MW: 443,42 g/mol
MDL Number: MFCD22666391
CAS Number: 1237744-13-6
https://us.vwr.com/stibo/web/std.lang.all/02/84/24140284.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR