CIQ ≥98% (by HPLC)

Synonyms: (3-Chlorophenyl) [3,4-dihydro-6,7-dimethoxy-1-[(4-methoxyphenoxy)methyl]-2(1H)-isoquinolinyl]methanone

B2005-5 B2005-25
76269-376EA 134.25 USD
76269-376 76269-378
CIQ ≥98% (by HPLC)
CIQ

CIQ is a subunit-selective potentiator of NMDA receptors containing the NR2C or NR2D subunit. Increases channel opening efficiency and enhances NMDA receptor responses.

Formula: C₂₆H₂₆ClNO₅
MW: 467,94 g/mol
CAS Number: 486427-17-2
https://us.vwr.com/stibo/web/std.lang.all/02/82/24140282.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR