Felbamate ≥98% (by HPLC)

Danger

Synonyms: ADD 030552-phenyl-1,3-propanediol 1,3-dicarbamate2-Phenylpropane-1,3-diyl dicarbamate

B2004-50 B2004-10
76269-374EA 484.51 USD
76269-374 76269-372
Felbamate ≥98% (by HPLC)
Felbamate

Felbamate is an antiepileptic drug that is effective against seizures in a variety of animal models. It displays inhibitory effects at NMDA and GABAA receptors and at voltage-gated channels.

Formula: C₁₁H₁₄N₂O₄
MW: 238,2 g/mol
MDL Number: MFCD00865296
CAS Number: 25451-15-4
https://us.vwr.com/stibo/web/std.lang.all/02/81/24140281.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR