Sapacitabine ≥98% (by HPLC)

Synonyms: N-(1-((2R,3S,4S,5R)-3-cyano-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)palmitamideCYC-682CS682CYC682

B2002-25 B2002-5
76269-364EA 262.69 USD
76269-364 76269-366
Sapacitabine ≥98% (by HPLC)
Sapacitabine

Sapacitabine is an orally bioavailable nucleoside analog prodrug that is potentially useful for treatment of hematologic malignancies and solid tumors. The active metabolite of sapacitabine, CNDAC (2'-C-cyano-2'-deoxy-1-β-D-arabino-pentofuranosylcytosine), displays the unique mechanism of action of causing single-strand breaks (SSBs) after incorporation into DNA, which are converted into double-strand breaks (DSBs) when cells enter a second S-phase. 

Formula: C₂₆H₄₂N₄O₅ 
MW: 490,64 g/mol
CAS Number: 151823-14-2 
https://us.vwr.com/stibo/web/std.lang.all/02/79/24140279.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR