PF-06291874 ≥98% (by HPLC)

Synonyms: N-[4-[(1S)-1-[3,5-Dimethyl-4-[4-(trifluoromethyl)-1H-pyrazol-1-yl]phenoxy]butyl]benzoyl]—alaninePF6291874.

B2001-5 B2001-25
76269-362EA 577.91 USD
76269-362 76269-360
PF-06291874 ≥98% (by HPLC)
PF-06291874

PF-06291874 is a glucagon receptor antagonist potentially useful for patients with type 2 diabetes mellitus.

Formula: C₂₆H₂₈F₃N₃O₄ 
MW: 503,52 g/mol
CAS Number: 1393124-08-7 
https://us.vwr.com/stibo/web/std.lang.all/02/78/24140278.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR