Saquinavir mesylate

Synonyms: (S)-N1-((2S,3R)-4-((3S,4AS,8aS)-3-(tert-butylcarbamoyl)octahydroisoquinolin-2(1H)-yl)-3-hydroxy-1-phenylbutan-2-yl)-2-(quinoline-2-carboxamido)succinamide methanesulfonate

B1999-50 B1999-10
76269-352EA 99.24 USD
76269-352 76269-354
Saquinavir mesylate
Saquinavir mesylate

Saquinavir mesylate is a HIV protease inhibitor (Kis = 0.12 and < 0.1 nM for HIV-1 and HIV-2 protease, respectively) that displays antiviral activity with low cytotoxicity.

Formula: C₃₈H₅₀N₆O₅·CH₄O₃S
MW: 766.95 g/mol
MDL Number: MFCD00944907
CAS Number: 149845-06-7
https://us.vwr.com/stibo/web/std.lang.all/02/76/24140276.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR