GBR-12935 ≥98% (by HPLC)
Synonyms:
1-(2 (Diphenylmethoxy)ethyl)-4-(3-phenylpropyl)piperazine
B1998-10
B1998-50
76269-348EA
134.25
USD
76269-348
76269-350
GBR-12935 ≥98% (by HPLC)
GBR-12935
GBR-12935 is a potent and selective dopamine reuptake inhibitor.
Formula:
C₂₈H₃₄N₂O MW: 414,59 g/mol |
CAS Number:
76778-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...