Chlorpromazine hydrochloride

Danger

Synonyms: 2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride

B1992-500 B1992-100
76269-274EA 262.69 USD
76269-274 76269-272
Chlorpromazine hydrochloride
Chlorpromazine hydrochloride

Chlorpromazine is a dopamine receptor antagonist; reduces neurotransmitter binding. Inhibits nitric oxide synthesis in mouse brain cytosol. Also acts as an antagonist of nAChR, histamine and 5-HT receptors. Displays growth-inhibitory effects on a variety of tumor cell lines. Exhibits antipsychotic activity.

Formula: C₁₇H₁₉ClN₂S
MW: 355.32 g/mol
MDL Number: MFCD00012654
CAS Number: 69-09-0
https://us.vwr.com/stibo/web/std.lang.all/02/70/24140270.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR