Valdecoxib ≥98% (by HPLC)

Danger

Synonyms: 4-(5-methyl-3-phenylisoxazol-4-yl)benzenesulphonamide

B1991-5 B1991-25
76269-228EA 145.93 USD
76269-228 76269-270
Valdecoxib ≥98% (by HPLC)
Valdecoxib

Valdecoxib is a non-steroidal anti-inflammatory drug (NSAID), that acts as a potent and selective inhibitor of cyclooxygenase-2 (COX-2) (IC₅₀ = 5 nM).

Formula: C₁₆H₁₄N₂O₃S
MW: 314,36 g/mol
MDL Number: MFCD00950568
CAS Number: 181695-72-7
https://us.vwr.com/stibo/web/std.lang.all/02/69/24140269.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR