Robenacoxib ≥98% (by HPLC)

Warning

Synonyms: 5-Ethyl-2-[(2,3,5,6-tetrafluorophenyl)amino]benzeneacetic acid

B1990-5 B1990-25
76269-224EA 110.91 USD
76269-224 76269-226
Robenacoxib ≥98% (by HPLC)
Robenacoxib

Robenacoxib is a non-steroidal anti-inflammatory drug (NSAID), that acts as a potent and selective inhibitor of cyclooxygenase-2 (COX-2).

Formula: C₁₆H₁₃F₄NO₂
MW: 327,27 g/mol
CAS Number: 220991-32-2
https://us.vwr.com/stibo/web/std.lang.all/02/68/24140268.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR