Parecoxib Sodium ≥98% (by HPLC)

Synonyms: Sodium ((4-(5-methyl-3-phenylisoxazol-4-yl)phenyl)sulfonyl)(propionyl)amideSodium ((4-(5-methyl-3-phenylisoxazol-4-yl)phenyl)sulphonyl)(propionyl)amideSC-69124ADynastat

B1989-10 B1989-50
76269-220EA 99.24 USD
76269-220 76269-222
Parecoxib Sodium ≥98% (by HPLC)
Parecoxib Sodium

Parecoxib sodium is an injectable prodrug of valdecoxib, that acts as a potent and selective inhibitor of cyclooxygenase-2 (COX-2). Useful for the short-term treatment of postoperative pain in adults. 

Formula: C₁₉H₁₇N₂NaO₄S
MW: 392,4 g/mol
MDL Number: MFCD04972608
CAS Number: 198470-85-8
https://us.vwr.com/stibo/web/std.lang.all/02/67/24140267.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR