Mavacoxib ≥98% (by HPLC)

Synonyms: 4-(5-(4-fluorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulfonamide PHA7395214-(5-(4-fluorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide

B1988-25 B1988-5
76269-216EA 134.25 USD
76269-216 76269-218
Mavacoxib ≥98% (by HPLC)
Mavacoxib

Mavacoxib is a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent and selective cyclooxygenase-2 (COX-2) inhibitor. Displays anti-inflammatory properties. Useful for treatment of dogs.

Formula: C₁₆H₁₁F₄N₃O₂S
MW: 385,34 g/mol
MDL Number: MFCD28138640
CAS Number: 170569-88-7 
https://us.vwr.com/stibo/web/std.lang.all/02/66/24140266.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR