Firocoxib ≥98% (by HPLC)

Danger

Synonyms: 3-(Cyclopropylmethoxy)-4-(4-methylsulphonylphenyl)-5,5-dimethylfuranone3-(Cyclopropylmethoxy)-4-(4-methylsulfonylphenyl)-5,5-dimethylfuranoneML1785713

B1986-5 B1986-25
76269-264EA 229.34 USD
76269-264 76269-266
Firocoxib ≥98% (by HPLC)
Firocoxib

Firocoxib is a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent and selective cyclooxygenase (COX)-2 inhibitor with an IC₅₀ of 0.13 uM. Displays 58-fold selectivity for COX-2 vs COX-1. A useful veterinary drug.

Formula: C₁₇H₂₀O₅S 
MW: 336,4 g/mol
CAS Number: 189954-96-9 
https://us.vwr.com/stibo/web/std.lang.all/02/64/24140264.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR