Cimicoxib ≥98% (by HPLC)

Danger

Synonyms: 4-[4-Chloro-5-(3-fluoro-4-methoxyphenyl)-1H-imidazol-1-yl]benzenesulphonamide4-[4-Chloro-5-(3-fluoro-4-methoxyphenyl)-1H-imidazol-1-yl]benzenesulfonamideUR-8880CimicoxibUR8880trade name: Cimalgex

B1984-25 B1984-5
76269-256EA 227.66 USD
76269-256 76269-258
Cimicoxib ≥98% (by HPLC)
Cimicoxib

Cimicoxib is an imidazole derivative and a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent and specific cyclooxygenase-2 (COX-2) inhibitor. Used in veterinary medicine to treat dogs for pain and inflammation associated with osteoarthritis and for the management of pain and inflammation associated with surgery. 

Formula: C₁₆H₁₃ClFN₃O₃S 
MW: 381,81 g/mol
CAS Number: 265114-23-6 
https://us.vwr.com/stibo/web/std.lang.all/02/62/24140262.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR