Etoricoxib ≥98% (by HPLC)

Danger

Synonyms: L-7914565-Chloro-6'-methyl-3-[4-(methylsulfonyl)phenyl]-2,3'-bipyridine5-Chloro-6'-methyl-3-[4-(methylsulphonyl)phenyl]-2,3'-bipyridineArcoxiaMK0663MK-0663

B1983-10 B1983-50
76269-252EA 99.22 USD
76269-252 76269-254
Etoricoxib ≥98% (by HPLC)
Etoricoxib

Etoricoxib is a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent and specific cyclooxygenase-2 (COX-2) inhibitor.

Formula: C₁₈H₁₅ClN₂O₂S 
MW: 358,84 g/mol
CAS Number: 202409-33-4 
https://us.vwr.com/stibo/web/std.lang.all/02/60/24140260.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR