Rofecoxib ≥98% (by HPLC)

Warning

Synonyms: MK-9664-(4-(Methylsulphonyl)phenyl)-3-phenylfuran-2(5H)-one4-(4-(Methylsulfonyl)phenyl)-3-phenylfuran-2(5H)-one

B1982-10 B1982-50
76269-248EA 99.24 USD
76269-248 76269-250
Rofecoxib ≥98% (by HPLC)
Rofecoxib

Rofecoxib is a non-steroidal anti-inflammatory drug (NSAID) that acts as a potent cyclooxygenase-2 (COX-2) inhibitor. It displays 1,000-fold selectivity for COX-2 over that of COX-1 (IC₅₀s = 26 nM and > 50 µM in human osteosarcoma cells).

Formula: C₁₇H₁₄O₄S 
MW: 314,36 g/mol
MDL Number: MFCD00935806
CAS Number: 162011-90-7
https://us.vwr.com/stibo/web/std.lang.all/02/59/24140259.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR